Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALG9 SIRNA Assays ›
          Gene Symbol:
          ALG9
          Gene Name:
          ALG9, alpha-1,2-mannosyltransferase
          Gene Aliases:
          CDG1L, DIBD1, GIKANIS, LOH11CR1J
          Chromosome Location:
          Chr.11: 111776096 - 111871581 on Build GRCh38
          UniGene ID:
          Hs.745155
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 130697

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ALG9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001077690.1 NP_001071158.1 6, 7 801
          NM_001077691.1 NP_001071159.1 6, 7 1078
          NM_001077692.1 NP_001071160.1 6, 7 1078
          NM_024740.2 NP_079016.2 6, 7 801
          XM_005277723.4 XP_005277780.1 6, 7 801
          XM_005277724.4 XP_005277781.1 6, 7 801
          XM_006718913.2 XP_006718976.1 6, 7 801
          XM_011542992.1 XP_011541294.1 6, 7 801
          XM_017018313.1 XP_016873802.1 6, 7 801
          XM_017018314.1 XP_016873803.1 6, 7 801
          XM_017018315.1 XP_016873804.1 6, 7 801
          XM_017018316.1 XP_016873805.1 6, 7 1098
          XM_017018317.1 XP_016873806.1 6, 7 1098
          XM_017018318.1 XP_016873807.1 6, 7 1105
          XM_017018319.1 XP_016873808.1 6, 7 935
          XM_017018320.1 XP_016873809.1 6, 7 805
          XM_017018321.1 XP_016873810.1 6, 7 941
          XM_017018322.1 XP_016873811.1 6, 7 714
          XM_017018323.1 XP_016873812.1 6, 7 1098
          XM_017018324.1 XP_016873813.1 6, 7 1085
          XM_017018325.1 XP_016873814.1 7, 8 1241
          XM_017018326.1 XP_016873815.1 6, 7 1102
          GenBank mRNA AF395532.1 - 6, 7 753
          AF454937.1 - 6, 7 712
          AK025498.1 - 6, 7 801
          AK172828.1 - 6, 7 662
          AK225261.1 - 6, 7 780
          AK298811.1 - 5, 6 641
          AK308025.1 - 5, 6 696
          AL136927.1 - 6, 7 724
          BC009255.2 - 6, 7 750 3029372
          KU178783.1 - 6, 7 703
          KU178784.1 - 6, 7 703

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          glycosyltransferase transferase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          dolichol-linked oligosaccharide biosynthetic process
          mannosylation
          alpha-1,2-mannosyltransferase activity
          mannosyltransferase activity
          dol-P-Man:Man(8)GlcNAc(2)-PP-Dol alpha-1,2-mannosyltransferase activity
          dol-P-Man:Man(6)GlcNAc(2)-PP-Dol alpha-1,2-mannosyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0