Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC38A1 SIRNA Assays ›
          Gene Symbol:
          SLC38A1
          Gene Name:
          solute carrier family 38 member 1
          Gene Aliases:
          ATA1, NAT2, SAT1, SNAT1
          Chromosome Location:
          Chr.12: 46183055 - 46269425 on Build GRCh38
          UniGene ID:
          Hs.533770
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 130945

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC38A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001077484.1 NP_001070952.1 12, 13 1671
          NM_001278387.1 NP_001265316.1 13, 14 2133
          NM_001278388.1 NP_001265317.1 11, 12 1895
          NM_001278389.1 NP_001265318.1 12, 13 1160
          NM_001278390.1 NP_001265319.1 12, 13 1873
          NM_030674.3 NP_109599.3 12, 13 2010
          XM_011538784.2 XP_011537086.1 12, 13 1395
          XM_011538786.2 XP_011537088.1 12, 13 1395
          XM_011538787.2 XP_011537089.1 12, 13 1395
          XM_017019990.1 XP_016875479.1 12, 13 1625
          GenBank mRNA AF247166.1 - 12, 13 1356
          AF271070.1 - 12, 13 1254
          AK027825.1 - 12, 13 1160
          AK074758.1 - 5, 6 427
          AK074797.1 - 9, 10 729
          AK074851.1 - 10, 11 872
          AK074949.1 - 10, 11 918
          AK225639.1 - 12, 13 1248
          AK225778.1 - 12, 13 1270
          AK308129.1 - 10, 11 958
          AK309410.1 - 12, 13 1587
          AK313731.1 - 10, 11 961
          BC010620.1 - 12, 13 1217 3871101
          BX648495.1 - 12, 13 1598
          BX648676.1 - 11, 12 1465

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          amino acid transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter uptake
          sodium ion transport
          amino acid transport
          asparagine transport
          glutamine transport
          neutral amino acid transport
          L-amino acid transport
          histidine transport
          L-histidine transmembrane transport
          sodium:amino acid symporter activity
          L-histidine transmembrane transporter activity
          protein binding
          amino acid transmembrane transporter activity
          neutral amino acid transmembrane transporter activity
          L-amino acid transmembrane transporter activity
          L-asparagine transmembrane transporter activity
          L-glutamine transmembrane transporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0