siRNA Details
Gene Symbol: ATM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000051.3 | NP_000042.3 | 4 | 662 | ||
XM_005271561.4 | XP_005271618.2 | 5 | 723 | |||
XM_005271562.4 | XP_005271619.2 | 4 | 796 | |||
XM_006718843.3 | XP_006718906.1 | 4 | 429 | |||
XM_011542840.2 | XP_011541142.1 | 5 | 517 | |||
XM_011542842.2 | XP_011541144.1 | 4 | 636 | |||
XM_011542843.2 | XP_011541145.1 | 4 | 1010 | |||
XM_011542844.2 | XP_011541146.1 | 4 | 634 | |||
XM_017017789.1 | XP_016873278.1 | 5 | 1220 | |||
XM_017017790.1 | XP_016873279.1 | 4 | 617 | |||
XM_017017791.1 | XP_016873280.1 | 4 | 1010 | |||
XM_017017792.1 | XP_016873281.1 | 4 | 1010 | |||
GenBank mRNA | AB209133.1 | - | 3 | 4800 | ||
BC061584.1 | - | 4 | 465 | |||
U33841.1 | - | 5 | 466 | |||
X91196.1 | - | 4 | 1010 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
