Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FIP1L1 SIRNA Assays ›
          Gene Symbol:
          FIP1L1
          Gene Name:
          factor interacting with PAPOLA and CPSF1
          Gene Aliases:
          FIP1, Rhe, hFip1
          Chromosome Location:
          Chr.4: 53377572 - 53462611 on Build GRCh38
          UniGene ID:
          Hs.555109
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 131014

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FIP1L1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134937.1 NP_001128409.1 11, 12 1156
          NM_001134938.1 NP_001128410.1 9, 10 979
          NM_030917.3 NP_112179.2 12, 13 1201
          XM_005265769.4 XP_005265826.1 12, 13 1268
          XM_005265773.4 XP_005265830.1 11, 12 1201
          XM_005265774.4 XP_005265831.1 11, 12 1140
          XM_005265778.4 XP_005265835.1 12, 13 1253
          XM_005265779.4 XP_005265836.1 12, 13 1208
          XM_005265781.4 XP_005265838.1 10, 11 1085
          XM_005265782.4 XP_005265839.1 10, 11 1031
          XM_017008662.1 XP_016864151.1 11, 12 1173
          XM_017008663.1 XP_016864152.1 11, 12 1102
          XM_017008664.1 XP_016864153.1 10, 11 1157
          XM_017008665.1 XP_016864154.1 11, 12 1100
          XM_017008666.1 XP_016864155.1 10, 11 1117
          XM_017008667.1 XP_016864156.1 10, 11 1056
          XM_017008668.1 XP_016864157.1 10, 11 1129
          XM_017008669.1 XP_016864158.1 9, 10 1002
          XM_017008670.1 XP_016864159.1 11, 12 1139
          XM_017008671.1 XP_016864160.1 10, 11 1031
          GenBank mRNA AK123992.1 - 3, 4 687
          AK295737.1 - 11, 12 1171
          AK300398.1 - 11, 12 903
          AK308145.1 - 12, 13 1011
          AL136910.1 - 9, 10 960
          AY366510.1 - 12, 13 1158
          BC011543.1 - 3, 4 219
          BC017724.1 - 11, 12 1138 3452247
          BC024016.1 - 10, 11 2243 4826276
          BC026724.1 - 1, 2 23
          BC052959.1 - 10, 11 2238 4838737
          BC110383.1 - 12, 13 1194 5165430

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          termination of RNA polymerase II transcription
          mRNA export from nucleus
          mRNA 3'-end processing
          protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0