Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FIP1L1 SIRNA Assays ›
Gene Symbol:
FIP1L1
Gene Name:
factor interacting with PAPOLA and CPSF1
Gene Aliases:
FIP1, Rhe, hFip1
Chromosome Location:
Chr.4: 53377572 - 53462611 on Build GRCh38
UniGene ID:
Hs.555109
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
131015
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FIP1L1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134937.1
NP_001128409.1
14
1377
NM_001134938.1
NP_001128410.1
11
1173
NM_030917.3
NP_112179.2
14
1395
XM_005265769.4
XP_005265826.1
15
1489
XM_005265773.4
XP_005265830.1
14
1422
XM_005265774.4
XP_005265831.1
13
1334
XM_005265778.4
XP_005265835.1
15
1474
XM_005265779.4
XP_005265836.1
14
1402
XM_005265781.4
XP_005265838.1
13
1306
XM_005265782.4
XP_005265839.1
12
1225
XM_017008662.1
XP_016864151.1
13
1367
XM_017008663.1
XP_016864152.1
14
1323
XM_017008664.1
XP_016864153.1
13
1378
XM_017008665.1
XP_016864154.1
13
1294
XM_017008666.1
XP_016864155.1
12
1311
XM_017008667.1
XP_016864156.1
13
1277
XM_017008668.1
XP_016864157.1
12
1323
XM_017008669.1
XP_016864158.1
12
1223
XM_017008670.1
XP_016864159.1
13
1333
XM_017008671.1
XP_016864160.1
13
1252
GenBank mRNA
AK090938.1
-
3
180
AK123992.1
-
5
881
AK295737.1
-
14
1392
AL136910.1
-
11
1154
AY366510.1
-
14
1352
BC011543.1
-
5
413
BC024016.1
-
12
2437
4826276
BC026724.1
-
3
217
BC052959.1
-
12
2432
4838737
BC110383.1
-
14
1388
5165430
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_024183, NM_001159573, NM_001159574, BC003263, AK049672, and others.
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
mRNA export from nucleus
mRNA 3'-end processing
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0