Search
Search

CDK2| Gene Symbol: CDK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290230.1 | NP_001277159.1 | 4 | 740 | ||
| NM_001798.4 | NP_001789.2 | 6 | 920 | |||
| NM_052827.3 | NP_439892.2 | 5 | 818 | |||
| XM_011537732.1 | XP_011536034.1 | 7 | 1064 | |||
| GenBank mRNA | AB012305.1 | - | 5 | 580 | ||
| AK291941.1 | - | 6 | 859 | |||
| AK293246.1 | - | 4 | 679 | |||
| BC003065.2 | - | 6 | 840 | 3504348 | ||
| BT006821.1 | - | 6 | 680 | |||
| M68520.1 | - | 6 | 870 | |||
| X61622.1 | - | 6 | 680 | |||
| X62071.1 | - | 6 | 850 | |||