Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DDR1 SIRNA Assays ›
          Gene Symbol:
          DDR1
          Gene Name:
          discoidin domain receptor tyrosine kinase 1
          Gene Aliases:
          CAK, CD167, DDR, EDDR1, HGK2, MCK10, NEP, NTRK4, PTK3, PTK3A, RTK6, TRKE
          Chromosome Location:
          Chr.6: 30880909 - 30900156 on Build GRCh38
          UniGene ID:
          Hs.631988
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 1316

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DDR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001202521.1 NP_001189450.1 3 314
          NM_001202522.1 NP_001189451.1 3 314
          NM_001202523.1 NP_001189452.1 4 566
          NM_001297652.1 NP_001284581.1 4 685
          NM_001297653.1 NP_001284582.1 4 487
          NM_001297654.1 NP_001284583.1 4 487
          NM_001954.4 NP_001945.3 6 605
          NM_013993.2 NP_054699.2 5 531
          NM_013994.2 NP_054700.2 3 314
          XM_006715185.2 XP_006715248.1 4 478
          XM_011514882.2 XP_011513184.1 4 478
          XM_011514883.1 XP_011513185.1 6 593
          XM_011514884.1 XP_011513186.1 6 669
          XM_011514885.1 XP_011513187.1 4 516
          XM_011514886.1 XP_011513188.1 4 381
          XM_011514887.1 XP_011513189.1 4 531
          XM_011514888.2 XP_011513190.1 4 478
          XM_017011268.1 XP_016866757.1 4 348
          XM_017011269.1 XP_016866758.1 4 531
          GenBank mRNA AB210021.1 - 5 426
          AK294793.1 - 4 552
          BC008716.2 - 6 605 2964574
          BC013400.2 - 6 605 2964574
          BC070070.1 - 4 376 5262509
          EU826613.1 - 3 272
          EU826614.1 - 3 272
          L20817.1 - 4 464
          L57508.1 - 4 464
          Z29093.1 - 3 608

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          regulation of cell-matrix adhesion
          cell adhesion
          embryo implantation
          lactation
          negative regulation of cell proliferation
          regulation of extracellular matrix disassembly
          smooth muscle cell migration
          extracellular matrix organization
          organ regeneration
          collagen-activated tyrosine kinase receptor signaling pathway
          peptidyl-tyrosine autophosphorylation
          ear development
          skin development
          wound healing, spreading of cells
          protein autophosphorylation
          branching involved in mammary gland duct morphogenesis
          mammary gland alveolus development
          smooth muscle cell-matrix adhesion
          transmembrane receptor protein tyrosine kinase activity
          protein binding
          collagen binding
          ATP binding
          protein tyrosine kinase collagen receptor activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0