Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREB3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052854.3 | NP_443086.1 | 1 | 560 | ||
XM_006718380.3 | XP_006718443.1 | 1 | 537 | |||
GenBank mRNA | AB063321.1 | - | 1 | 181 | ||
AK314072.1 | - | 1 | 538 | |||
BC014097.1 | - | 1 | 203 | 4546950 | ||
BC015781.1 | - | 1 | 189 | 4856946 | ||
BQ953032.1 | - | 1 | 520 | |||
CF131922.1 | - | 1 | 556 |