Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282161.1 | NP_001269090.1 | 6 | 1732 | ||
NM_003336.3 | NP_003327.2 | 6 | 1739 | |||
NM_181762.2 | NP_861427.1 | 5 | 1649 | |||
GenBank mRNA | AK223045.1 | - | 6 | 1641 | ||
AK223064.1 | - | 6 | 1638 | |||
BC042021.1 | - | 3 | 2516 | |||
M74524.1 | - | 6 | 1614 |