Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128324.2 | NP_001121796.1 | 4 | 570 | ||
NM_001282905.1 | NP_001269834.1 | 5 | 1020 | |||
NM_001282906.1 | NP_001269835.1 | 5 | 672 | |||
NM_174890.3 | NP_777550.2 | 4 | 918 | |||
NR_104256.1 | 4 | 570 | ||||
XM_005271842.2 | XP_005271899.1 | 3 | 443 | |||
XM_005271843.2 | XP_005271900.1 | 3 | 610 | |||
XM_017016932.1 | XP_016872421.1 | 4 | 662 | |||
XM_017016933.1 | XP_016872422.1 | 4 | 529 | |||
XM_017016934.1 | XP_016872423.1 | 4 | 582 | |||
XM_017016935.1 | XP_016872424.1 | 3 | 332 | |||
XM_017016940.1 | XP_016872429.1 | 5 | 1078 | |||
XM_017016941.1 | XP_016872430.1 | 5 | 621 | |||
XM_017016942.1 | XP_016872431.1 | 3 | 610 | |||
XM_017016943.1 | XP_016872432.1 | 3 | 886 | |||
XM_017016944.1 | XP_016872433.1 | 3 | 443 | |||
XM_017016946.1 | XP_016872435.1 | 4 | 529 | |||
GenBank mRNA | AF311324.1 | - | 4 | 534 | ||
AK090704.1 | - | 5 | 744 | |||
AK097924.1 | - | 4 | 537 | |||
AK292420.1 | - | 5 | 621 | |||
AK292469.1 | - | 4 | 519 | |||
AK314395.1 | - | 4 | 530 | |||
BC045587.1 | - | 4 | 570 | 5269483 | ||
BC048968.1 | - | 5 | 562 | 5272133 |