Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other WDR20 SIRNA Assays ›
Gene Symbol:
WDR20
Gene Name:
WD repeat domain 20
Gene Aliases:
DMR
Chromosome Location:
Chr.14: 102139518 - 102224686 on Build GRCh38
UniGene ID:
Hs.36859
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
131867
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: WDR20
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242414.1
NP_001229343.1
3
543
NM_001242416.1
NP_001229345.1
2
530
NM_001242417.1
NP_001229346.1
4
806
NM_001242418.1
NP_001229347.1
3
749
NM_144574.3
NP_653175.2
3
713
NM_181291.2
NP_851808.1
3
713
NM_181308.2
NP_851825.1
2
530
XM_006720308.1
XP_006720371.1
2
522
XM_006720309.1
XP_006720372.1
3
496
XM_006720310.1
XP_006720373.1
3
457
XM_006720311.1
XP_006720374.1
2
286
XM_011537335.2
XP_011535637.1
4
825
XM_011537336.2
XP_011535638.1
4
825
XM_011537337.2
XP_011535639.1
4
825
XM_011537338.1
XP_011535640.1
4
772
XM_011537339.2
XP_011535641.1
4
825
XM_011537340.2
XP_011535642.1
4
825
XM_011537341.2
XP_011535643.1
3
731
XM_011537343.1
XP_011535645.1
5
800
XM_011537344.2
XP_011535646.1
4
718
XM_011537345.1
XP_011535647.1
3
496
XM_011537346.1
XP_011535648.1
3
522
XM_011537347.1
XP_011535649.1
4
520
XM_017021766.1
XP_016877255.1
4
772
XM_017021767.1
XP_016877256.1
3
731
XM_017021768.1
XP_016877257.1
3
746
XM_017021770.1
XP_016877259.1
3
746
XM_017021771.1
XP_016877260.1
4
781
XM_017021772.1
XP_016877261.1
4
534
XM_017021773.1
XP_016877262.1
5
627
XM_017021774.1
XP_016877263.1
4
576
XM_017021775.1
XP_016877264.1
4
582
XM_017021776.1
XP_016877265.1
3
496
XM_017021777.1
XP_016877266.1
3
464
XM_017021778.1
XP_016877267.1
4
576
XM_017021779.1
XP_016877268.1
2
286
XM_017021780.1
XP_016877269.1
4
697
XM_017021785.1
XP_016877274.1
4
772
GenBank mRNA
AB209148.1
-
1
4802
AF327354.1
-
3
638
AK056330.1
-
3
455
AK090978.1
-
3
484
AK225852.1
-
2
489
AK297727.1
-
4
744
AL832242.1
-
3
482
AL832500.1
-
3
688
BC028387.2
-
3
672
4823662
BX248274.1
-
2
432
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_027149, BC026761, AK136417, AK052481, AK145389, and others.
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0