Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRPM3 SIRNA Assays ›
Gene Symbol:
TRPM3
Gene Name:
transient receptor potential cation channel subfamily M member 3
Gene Aliases:
GON-2, LTRPC3, MLSN2
Chromosome Location:
Chr.9: 70529063 - 71446989 on Build GRCh38
UniGene ID:
Hs.47288
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
131901
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRPM3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001007470.1
NP_001007471.1
8
1000
NM_001007471.2
NP_001007472.2
7
1273
NM_020952.4
NP_066003.3
7
925
NM_024971.5
NP_079247.5
7
925
NM_206944.3
NP_996827.3
7
925
NM_206945.3
NP_996828.3
7
925
NM_206946.3
NP_996829.3
8
1000
NM_206947.3
NP_996830.3
8
1000
NM_206948.2
NP_996831.1
7
925
XM_011519035.2
XP_011517337.1
8
1225
XM_011519036.2
XP_011517338.1
8
4150
XM_011519037.2
XP_011517339.1
8
4136
XM_011519038.2
XP_011517340.1
8
4133
XM_011519039.2
XP_011517341.1
8
4134
XM_011519040.2
XP_011517342.1
7
1156
XM_011519041.2
XP_011517343.1
7
4058
XM_011519042.2
XP_011517344.1
7
1156
XM_011519043.2
XP_011517345.1
7
1189
XM_011519044.2
XP_011517346.1
7
4059
XM_011519045.2
XP_011517347.1
7
1150
XM_011519046.2
XP_011517348.1
7
2853
XM_011519047.2
XP_011517349.1
7
2823
XM_017015141.1
XP_016870630.1
8
1225
XM_017015142.1
XP_016870631.1
8
1225
XM_017015143.1
XP_016870632.1
8
1225
XM_017015144.1
XP_016870633.1
8
991
XM_017015145.1
XP_016870634.1
8
1669
XM_017015146.1
XP_016870635.1
8
1067
XM_017015147.1
XP_016870636.1
8
3102
XM_017015148.1
XP_016870637.1
8
1225
XM_017015149.1
XP_016870638.1
8
1225
XM_017015150.1
XP_016870639.1
8
1225
XM_017015151.1
XP_016870640.1
7
1168
XM_017015152.1
XP_016870641.1
8
2903
XM_017015153.1
XP_016870642.1
7
1160
XM_017015154.1
XP_016870643.1
8
1371
XM_017015155.1
XP_016870644.1
7
2825
XM_017015156.1
XP_016870645.1
3
348
XM_017015157.1
XP_016870646.1
7
2823
XM_017015158.1
XP_016870647.1
7
2821
XM_017015159.1
XP_016870648.1
7
916
XM_017015160.1
XP_016870649.1
7
1006
XM_017015161.1
XP_016870650.1
8
1225
GenBank mRNA
AB099661.1
-
4
570
AB099662.1
-
5
645
AB099663.1
-
4
570
AB099664.1
-
5
645
AB099665.1
-
4
570
AF325212.1
-
7
896
AF536748.1
-
4
570
AF536749.1
-
4
570
AF536750.1
-
4
570
AF536751.1
-
4
570
AF536752.1
-
4
570
AF536753.1
-
5
645
AJ505025.1
-
7
1273
AJ505026.1
-
7
1273
AK225732.1
-
7
916
AK308682.1
-
7
1101
BC022454.2
-
7
930
BC067733.1
-
8
962
BC094699.1
-
7
919
BC121821.1
-
5
771
BC134414.1
-
5
774
BC142972.1
-
7
942
KF987075.1
-
7
981
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
transporter
Gene Ontology Categories:
Function(s)
Process(es)
cation transport
detection of temperature stimulus
sensory perception of temperature stimulus
protein tetramerization
calcium ion transmembrane transport
calcium activated cation channel activity
cation channel activity
calcium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-c4z4p:80/10.239.161.116:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0