Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007259.2 | NP_001007260.1 | 4, 5 | 386 | ||
NM_001007261.2 | NP_001007262.1 | 5, 6 | 480 | |||
NM_001286752.1 | NP_001273681.1 | 5, 6 | 869 | |||
NM_017438.4 | NP_059134.1 | 4, 5 | 726 | |||
XM_011529636.1 | XP_011527938.1 | 4, 5 | 775 | |||
XM_011529637.1 | XP_011527939.1 | 4, 5 | 840 | |||
XM_011529638.2 | XP_011527940.1 | 4, 5 | 748 | |||
XM_011529639.1 | XP_011527941.1 | 4, 5 | 526 | |||
XM_011529640.2 | XP_011527942.1 | 4, 5 | 791 | |||
XM_011529642.1 | XP_011527944.1 | 5, 6 | 869 | |||
XM_011529643.1 | XP_011527945.1 | 5, 6 | 820 | |||
XM_011529644.1 | XP_011527946.1 | 5, 6 | 381 | |||
XM_017028403.1 | XP_016883892.1 | 4, 5 | 317 | |||
XM_017028404.1 | XP_016883893.1 | 5, 6 | 425 | |||
XM_017028405.1 | XP_016883894.1 | 4, 5 | 726 | |||
GenBank mRNA | AB004848.1 | - | 5, 6 | 433 | ||
AB004852.1 | - | 3, 4 | 257 | |||
AB004853.1 | - | 5, 6 | 429 | |||
AK001660.1 | - | 4, 5 | 293 | |||
AK095215.1 | - | 3, 4 | 1593 | |||
BC002898.2 | - | 4, 5 | 325 | 3940849 | ||
BC036556.2 | - | 5, 6 | 543 | 5266627 | ||
BF195582.1 | - | 4, 5 | 386 |