Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017589.3 | NP_060059.1 | 3 | 486 | ||
XM_011542876.2 | XP_011541178.1 | 3 | 520 | |||
XM_011542878.2 | XP_011541180.1 | 3 | 520 | |||
XM_011542879.2 | XP_011541181.1 | 3 | 520 | |||
XM_011542880.2 | XP_011541182.1 | 3 | 520 | |||
GenBank mRNA | AJ271351.1 | - | 2 | 291 | ||
BC031045.1 | - | 3 | 472 | 5271706 |