Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCOLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253693.1 | NP_001240622.1 | 10 | 1245 | ||
NM_018298.10 | NP_060768.8 | 11 | 1413 | |||
XM_005271003.1 | XP_005271060.1 | 12 | 1810 | |||
XM_006710750.1 | XP_006710813.1 | 12 | 1629 | |||
XM_011541739.2 | XP_011540041.2 | 11 | 1337 | |||
XM_011541740.2 | XP_011540042.1 | 10 | 1171 | |||
XM_011541742.1 | XP_011540044.1 | 12 | 1547 | |||
GenBank mRNA | AF475085.1 | - | 11 | 1319 | ||
AK001868.1 | - | 10 | 1151 | |||
AK093948.1 | - | 2 | 1290 | |||
AK292206.1 | - | 11 | 1345 | |||
AK314354.1 | - | 11 | 1373 |