Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MBNL3 SIRNA Assays ›
Gene Symbol:
MBNL3
Gene Name:
muscleblind like splicing regulator 3
Gene Aliases:
CHCR, MBLX, MBLX39, MBXL
Chromosome Location:
Chr.X: 132369314 - 132490030 on Build GRCh38
UniGene ID:
Hs.105134
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
132493
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MBNL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170701.1
NP_001164172.1
2
224
NM_001170702.1
NP_001164173.1
2
224
NM_001170703.1
NP_001164174.1
2
157
NM_001170704.1
NP_001164175.1
3
424
NM_018388.3
NP_060858.2
2
387
NM_133486.2
NP_597846.1
2
387
XM_005262431.3
XP_005262488.1
3
1199
XM_005262434.3
XP_005262491.1
3
1194
XM_005262436.3
XP_005262493.1
2
1087
XM_005262437.3
XP_005262494.1
2
225
XM_005262438.3
XP_005262495.1
2
225
XM_005262439.3
XP_005262496.1
2
157
XM_005262440.3
XP_005262497.1
4
1244
XM_005262441.3
XP_005262498.1
2
348
XM_005262442.3
XP_005262499.1
2
319
XM_017029632.1
XP_016885121.1
3
8629
XM_017029633.1
XP_016885122.1
4
1364
XM_017029634.1
XP_016885123.1
3
8629
XM_017029635.1
XP_016885124.1
3
8629
XM_017029636.1
XP_016885125.1
3
1128
XM_017029637.1
XP_016885126.1
3
8629
XM_017029638.1
XP_016885127.1
4
1262
XM_017029639.1
XP_016885128.1
2
337
GenBank mRNA
AB077698.1
-
2
387
AB077699.1
-
2
387
AF467070.1
-
2
387
AF491305.1
-
2
308
AJ427918.1
-
2
387
AJ427919.1
-
2
387
AK002178.1
-
2
193
AK300248.1
-
2
156
AK307194.1
-
4
1241
AL133625.1
-
1
132
AY072692.1
-
2
387
AY372211.1
-
2
158
BC042090.1
-
3
423
5761647
BC074775.2
-
2
350
30915333
BC074776.2
-
2
350
30915555
BU569439.1
-
2
227
DB023330.1
-
2
157
DC380677.1
-
2
353
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106949, XM_008773562, XM_008773558, XM_008773556, XM_008773559, and others.
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
multicellular organism development
RNA splicing
regulation of RNA splicing
negative regulation of myoblast differentiation
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-29whj:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0