Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TSNAXIP1 SIRNA Assays ›
Gene Symbol:
TSNAXIP1
Gene Name:
translin associated factor X interacting protein 1
Gene Aliases:
TXI1
Chromosome Location:
Chr.16: 67806573 - 67828068 on Build GRCh38
UniGene ID:
Hs.632212
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
132522
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TSNAXIP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001288990.1
NP_001275919.1
15
2134
NM_001288991.1
NP_001275920.1
13
1927
NM_001288992.1
NP_001275921.1
11
1880
NM_001288993.1
NP_001275922.1
12
2007
NM_001288994.1
NP_001275923.1
10
1683
NM_018430.3
NP_060900.2
15
2323
XM_005256049.4
XP_005256106.1
15
2328
XM_011523230.2
XP_011521532.1
14
2535
XM_011523231.2
XP_011521533.1
14
2514
XM_011523232.2
XP_011521534.1
15
2487
XM_011523233.2
XP_011521535.1
15
2460
XM_011523234.2
XP_011521536.1
14
2403
XM_011523235.1
XP_011521537.1
14
2170
XM_011523236.2
XP_011521538.1
12
1793
XM_011523237.2
XP_011521539.1
12
2295
XM_011523238.1
XP_011521540.1
11
1708
XM_011523239.1
XP_011521541.1
12
1709
XM_011523240.1
XP_011521542.1
13
1895
XM_011523241.1
XP_011521543.1
12
1983
XM_011523242.1
XP_011521544.1
11
1836
XM_011523243.1
XP_011521545.1
11
1790
XM_017023445.1
XP_016878934.1
12
1772
XM_017023446.1
XP_016878935.1
12
1639
XM_017023447.1
XP_016878936.1
12
1661
XM_017023448.1
XP_016878937.1
12
1612
XM_017023449.1
XP_016878938.1
13
1586
XM_017023450.1
XP_016878939.1
14
1799
XM_017023451.1
XP_016878940.1
14
1985
XM_017023452.1
XP_016878941.1
11
1815
XM_017023453.1
XP_016878942.1
13
1887
XM_017023454.1
XP_016878943.1
12
1740
XM_017023455.1
XP_016878944.1
12
1629
GenBank mRNA
AF132730.1
-
12
1811
AK027245.1
-
15
2041
AK301920.1
-
15
1907
AK303841.1
-
13
1717
BC111018.1
-
15
2077
5168384
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
spermatogenesis
biological_process
cell differentiation
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0