Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TSNAXIP1 SIRNA Assays ›
          Gene Symbol:
          TSNAXIP1
          Gene Name:
          translin associated factor X interacting protein 1
          Gene Aliases:
          TXI1
          Chromosome Location:
          Chr.16: 67806573 - 67828068 on Build GRCh38
          UniGene ID:
          Hs.632212
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 132523

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TSNAXIP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001288990.1 NP_001275919.1 15, 16 2164
          NM_001288991.1 NP_001275920.1 13, 14 1957
          NM_001288992.1 NP_001275921.1 11, 12 1910
          NM_001288993.1 NP_001275922.1 12, 13 2037
          NM_001288994.1 NP_001275923.1 10, 11 1713
          NM_018430.3 NP_060900.2 15, 16 2353
          XM_005256049.4 XP_005256106.1 15, 16 2358
          XM_011523230.2 XP_011521532.1 14, 15 2565
          XM_011523231.2 XP_011521533.1 14, 15 2544
          XM_011523232.2 XP_011521534.1 15, 16 2517
          XM_011523233.2 XP_011521535.1 15, 16 2490
          XM_011523234.2 XP_011521536.1 14, 15 2433
          XM_011523235.1 XP_011521537.1 14, 15 2200
          XM_011523236.2 XP_011521538.1 12, 13 1823
          XM_011523237.2 XP_011521539.1 12, 13 2325
          XM_011523238.1 XP_011521540.1 11, 12 1738
          XM_011523239.1 XP_011521541.1 12, 13 1739
          XM_011523240.1 XP_011521542.1 13, 14 1925
          XM_011523241.1 XP_011521543.1 12, 13 2013
          XM_011523242.1 XP_011521544.1 11, 12 1866
          XM_011523243.1 XP_011521545.1 11, 12 1820
          XM_017023445.1 XP_016878934.1 12, 13 1802
          XM_017023446.1 XP_016878935.1 12, 13 1669
          XM_017023447.1 XP_016878936.1 12, 13 1691
          XM_017023448.1 XP_016878937.1 12, 13 1642
          XM_017023449.1 XP_016878938.1 13, 14 1616
          XM_017023450.1 XP_016878939.1 14, 15 1829
          XM_017023451.1 XP_016878940.1 14, 15 2015
          XM_017023452.1 XP_016878941.1 11, 12 1845
          XM_017023453.1 XP_016878942.1 13, 14 1917
          XM_017023454.1 XP_016878943.1 12, 13 1770
          XM_017023455.1 XP_016878944.1 12, 13 1659
          GenBank mRNA AF132730.1 - 12, 13 1841
          AK027245.1 - 15, 16 2071
          AK301920.1 - 15, 16 1937
          AK303841.1 - 13, 14 1747
          BC111018.1 - 15, 16 2107 5168384

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          spermatogenesis
          biological_process
          cell differentiation
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0