Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320041.1 | NP_001306970.1 | 4 | 469 | ||
NM_018456.5 | NP_060926.2 | 5 | 606 | |||
XM_005247618.3 | XP_005247675.1 | 6 | 756 | |||
XM_017006861.1 | XP_016862350.1 | 6 | 478 | |||
XM_017006862.1 | XP_016862351.1 | 4 | 378 | |||
XM_017006863.1 | XP_016862352.1 | 5 | 549 | |||
GenBank mRNA | AF217516.1 | - | 4 | 467 | ||
AF517829.1 | - | 5 | 551 | |||
AK301560.1 | - | 5 | 549 | |||
AY049020.1 | - | 5 | 571 | |||
BC014209.2 | - | 5 | 566 | 4765823 |