Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHGC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018928.2 | NP_061751.1 | 1 | 1648 | ||
NM_032406.1 | NP_115782.1 | 1 | 1648 | |||
GenBank mRNA | AF152338.1 | - | 1 | 1648 | ||
AF152525.1 | - | 1 | 1648 | |||
BC101035.1 | - | 1 | 1648 | 40007872 | ||
BC101036.1 | - | 1 | 1645 | |||
BC101038.2 | - | 1 | 1648 | 40007875 |