Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BBX SIRNA Assays ›
          Gene Symbol:
          BBX
          Gene Name:
          BBX, HMG-box containing
          Gene Aliases:
          ARTC1, HBP2, HSPC339, MDS001
          Chromosome Location:
          Chr.3: 107522936 - 107811329 on Build GRCh38
          UniGene ID:
          Hs.124366
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 133038

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BBX
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001142568.2 NP_001136040.1 9 1124
          NM_001276286.1 NP_001263215.1 9 1124
          NM_020235.6 NP_064620.2 9 1124
          XM_005247642.3 XP_005247699.1 9 1040
          XM_005247643.3 XP_005247700.1 9 1052
          XM_005247644.3 XP_005247701.1 10 1215
          XM_011512999.2 XP_011511301.1 9 1124
          XM_011513000.1 XP_011511302.1 9 1252
          XM_011513001.1 XP_011511303.1 9 1092
          XM_011513002.2 XP_011511304.1 9 1124
          XM_011513003.2 XP_011511305.1 9 1124
          XM_011513004.2 XP_011511306.1 9 1252
          XM_011513005.2 XP_011511307.1 9 1124
          XM_011513006.2 XP_011511308.1 9 1124
          XM_011513007.2 XP_011511309.1 9 1124
          XM_011513010.2 XP_011511312.1 9 1124
          XM_017006881.1 XP_016862370.1 11 1265
          XM_017006882.1 XP_016862371.1 10 1215
          XM_017006883.1 XP_016862372.1 9 1124
          GenBank mRNA AF161457.1 - 9 1104
          AF276948.1 - 6 822
          AF276949.1 - 9 1055
          AF283775.2 - 9 1118
          AF454941.1 - 9 1117
          AF454942.1 - 7 997
          AK057010.1 - 9 1070
          AK291757.1 - 9 1070
          AK307668.1 - 8 1164
          AK314434.1 - 9 1075
          AL136769.1 - 7 997
          BC110903.1 - 7 1034
          BC131718.1 - 9 1017 40123389

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          bone development
          DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-hmbwg:80/100.66.77.66:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline