Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318785.1 | NP_001305714.1 | 19 | 1968 | ||
NM_020320.4 | NP_064716.2 | 20 | 1803 | |||
NR_134857.1 | 20 | 1799 | ||||
XM_017011079.1 | XP_016866568.1 | 20 | 1969 | |||
XM_017011080.1 | XP_016866569.1 | 21 | 2131 | |||
XM_017011081.1 | XP_016866570.1 | 19 | 1734 | |||
GenBank mRNA | AK023550.1 | - | 20 | 1746 | ||
AK093934.1 | - | 20 | 2176 | |||
BC010420.1 | - | 20 | 1774 | 3613406 | ||
BC105784.1 | - | 19 | 1907 | |||
CB177631.1 | - | 1 | 76 |