Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003619.3 | NP_003610.2 | 2 | 596 | ||
XM_005263318.4 | XP_005263375.1 | 2 | 869 | |||
XM_011532387.2 | XP_011530689.1 | 2 | 871 | |||
GenBank mRNA | AJ001531.1 | - | 2 | 589 | ||
BC007761.2 | - | 2 | 596 | 4126901 | ||
BG386264.1 | - | 2 | 450 |