Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282660.1 | NP_001269589.1 | 9 | 1356 | ||
NM_001282661.1 | NP_001269590.1 | 11 | 1618 | |||
NM_001282662.1 | NP_001269591.1 | 12 | 1768 | |||
NM_001318139.1 | NP_001305068.1 | 10 | 1493 | |||
NM_020831.4 | NP_065882.1 | 12 | 1768 | |||
XM_005261692.1 | XP_005261749.1 | 11 | 1573 | |||
XM_005261694.1 | XP_005261751.1 | 11 | 1663 | |||
XM_011530283.1 | XP_011528585.1 | 13 | 1876 | |||
XM_011530285.1 | XP_011528587.1 | 13 | 1787 | |||
XM_011530287.1 | XP_011528589.1 | 5 | 755 | |||
XM_017028888.1 | XP_016884377.1 | 12 | 1851 | |||
XM_017028889.1 | XP_016884378.1 | 8 | 2851 | |||
GenBank mRNA | AB037859.2 | - | 12 | 1740 | ||
AF368061.1 | - | 12 | 1693 | |||
AJ297257.1 | - | 9 | 1350 | |||
AJ297258.1 | - | 12 | 1750 | |||
AL713710.1 | - | 4 | 563 | |||
BC115039.1 | - | 12 | 1734 | 5551767 | ||
BC139730.1 | - | 10 | 1407 | |||
CR456522.1 | - | 9 | 1167 | |||
HG764171.1 | - | 10 | 1493 |