Search
Search

RDH14
NT5C1B-RDH14| Gene Symbol: RDH14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020905.3 | NP_065956.1 | 2 | 851 | ||
| GenBank mRNA | AF237952.1 | - | 2 | 793 | ||
| AK023625.1 | - | 1 | 1178 | |||
| AK314994.1 | - | 2 | 811 | |||
| AY358511.1 | - | 2 | 794 | |||
| BC009830.2 | - | 2 | 751 | 4053893 | ||
| Gene Symbol: NT5C1B-RDH14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199103.1 | NP_001186032.1 | 9 | 1784 | ||
| NM_001199104.1 | NP_001186033.1 | 11 | 2233 | |||