Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PELI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021255.2 | NP_067078.1 | 6 | 3543 | ||
XM_006720211.3 | XP_006720274.1 | 5 | 5678 | |||
XM_011536990.2 | XP_011535292.1 | 7 | 3253 | |||
XM_011536992.2 | XP_011535294.1 | 6 | 3306 | |||
XM_017021478.1 | XP_016876967.1 | 6 | 3935 | |||
XM_017021479.1 | XP_016876968.1 | 7 | 12485 | |||
GenBank mRNA | AF302502.1 | - | 6 | 3436 | ||
AL133605.1 | - | 1 | 2147 | |||
AL709755.1 | - | 1 | 226 |