Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C21orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286463.1 | NP_001273392.1 | 7 | 1487 | ||
NM_001286476.1 | NP_001273405.1 | 9 | 2466 | |||
NM_001286477.1 | NP_001273406.1 | 8 | 2128 | |||
NM_058180.4 | NP_478060.2 | 8 | 2500 | |||
XM_005261148.4 | XP_005261205.1 | 9 | 2203 | |||
XM_011529621.2 | XP_011527923.1 | 8 | 2483 | |||
XM_011529623.2 | XP_011527925.1 | 8 | 4488 | |||
XM_011529624.2 | XP_011527926.1 | 8 | 1363 | |||
XM_011529625.2 | XP_011527927.1 | 8 | 1637 | |||
XM_011529626.2 | XP_011527928.1 | 9 | 4834 | |||
XM_011529627.2 | XP_011527929.1 | 9 | 4914 | |||
XM_017028388.1 | XP_016883877.1 | 9 | 4665 | |||
XM_017028389.1 | XP_016883878.1 | 10 | 4551 | |||
XM_017028390.1 | XP_016883879.1 | 9 | 2832 | |||
XM_017028391.1 | XP_016883880.1 | 8 | 2655 | |||
XM_017028392.1 | XP_016883881.1 | 10 | 1852 | |||
XM_017028393.1 | XP_016883882.1 | 9 | 1651 | |||
GenBank mRNA | AK056387.1 | - | 8 | 2473 | ||
AK096641.1 | - | 7 | 2404 | |||
AK098098.1 | - | 7 | 1424 | |||
BC026042.1 | - | 9 | 2441 | |||
BM475055.1 | - | 1 | 240 |