Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APTX SIRNA Assays ›
Gene Symbol:
APTX
Gene Name:
aprataxin
Gene Aliases:
AOA, AOA1, AXA1, EAOH, EOAHA, FHA-HIT
Chromosome Location:
Chr.9: 32883872 - 33025129 on Build GRCh38
UniGene ID:
Hs.20158
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
133724
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: APTX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001195248.1
NP_001182177.1
6
663
NM_001195249.1
NP_001182178.1
6
832
NM_001195250.1
NP_001182179.1
5
501
NM_001195251.1
NP_001182180.1
7
769
NM_001195252.1
NP_001182181.1
6
447
NM_001195254.1
NP_001182183.1
5
523
NM_175069.2
NP_778239.1
6
663
NM_175073.2
NP_778243.1
7
769
NR_036576.1
6
658
NR_036577.1
6
548
NR_036578.1
6
680
NR_036579.1
6
827
XM_006716791.3
XP_006716854.1
6
671
XM_006716792.2
XP_006716855.1
6
689
XM_011517938.1
XP_011516240.1
5
647
XM_011517939.2
XP_011516241.1
5
602
XM_017014831.1
XP_016870320.1
6
663
XM_017014832.1
XP_016870321.1
6
663
XM_017014833.1
XP_016870322.1
6
663
XM_017014834.1
XP_016870323.1
6
663
XM_017014835.1
XP_016870324.1
6
663
XM_017014836.1
XP_016870325.1
6
663
XM_017014837.1
XP_016870326.1
5
752
XM_017014838.1
XP_016870327.1
6
645
GenBank mRNA
AB603741.1
-
5
559
AJ565850.1
-
6
521
AJ565851.1
-
6
526
AJ565852.1
-
6
636
AJ565853.1
-
5
639
AJ565854.1
-
5
644
AJ565855.1
-
4
482
AJ575566.1
-
5
524
AK000164.1
-
5
666
AK055672.1
-
6
650
AK131046.1
-
6
615
AY040777.1
-
6
638
AY208829.1
-
6
638
AY208830.1
-
5
501
AY208831.1
-
6
413
AY208832.1
-
5
476
AY208833.1
-
5
476
AY208834.1
-
5
577
AY208835.1
-
6
618
AY208836.1
-
6
778
AY208837.1
-
7
735
AY208838.1
-
5
594
AY208839.1
-
6
772
AY208840.1
-
6
635
AY208841.1
-
5
472
AY208842.1
-
3
272
AY302067.1
-
5
781
AY302068.1
-
5
776
AY302069.1
-
5
666
AY302070.1
-
5
661
BC001628.1
-
5
486
BC032650.1
-
6
704
5505120
BC068107.1
-
7
744
BC104881.1
-
6
677
8143884
BX538161.1
-
7
748
DB042208.1
-
5
523
Back To Top
More Information
Panther Classification:
Molecular Function -
damaged DNA-binding protein
Gene Ontology Categories:
Function(s)
Process(es)
single strand break repair
DNA ligation
cellular response to DNA damage stimulus
dephosphorylation
regulation of protein stability
response to hydrogen peroxide
nucleic acid phosphodiester bond hydrolysis
chromatin binding
damaged DNA binding
double-stranded DNA binding
double-stranded RNA binding
protein binding
phosphoglycolate phosphatase activity
DNA 5'-adenosine monophosphate hydrolase activity
metal ion binding
protein N-terminus binding
phosphoprotein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0