Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199534.1 | NP_001186463.1 | 4 | 683 | ||
NM_018840.4 | NP_061328.1 | 4 | 655 | |||
NM_199483.2 | NP_955777.1 | 3 | 525 | |||
NR_026562.3 | 5 | 766 | ||||
GenBank mRNA | AF112213.1 | - | 4 | 619 | ||
AF274936.1 | - | 4 | 559 | |||
AF274948.1 | - | 4 | 468 | |||
BC001871.1 | - | 4 | 600 | 3546888 | ||
BC004446.1 | - | 5 | 691 | 2819807 | ||
BI223439.1 | - | 4 | 660 | |||
BU189639.1 | - | 3 | 437 | |||
BU194101.1 | - | 452 | ||||
BU848918.1 | - | 4 | 614 |
Gene Symbol: TGIF2-C20orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199535.1 | NP_001186464.1 | 5 | 631 |