Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DLL1 SIRNA Assays ›
          Gene Symbol:
          DLL1
          Gene Name:
          delta like canonical Notch ligand 1
          Gene Aliases:
          DELTA1, DL1, Delta
          Chromosome Location:
          Chr.6: 170282200 - 170291075 on Build GRCh38
          UniGene ID:
          Hs.379912
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 133773

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DLL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005618.3 NP_005609.3 4 984
          XM_005266934.3 XP_005266991.1 4 1450
          GenBank mRNA AF196571.1 - 4 832
          AK314234.1 - 4 856
          AY358892.1 - 4 551
          DR000029.1 - 4 603
          EU927387.1 - 4 675

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cell fate determination
          somitogenesis
          somite specification
          heart looping
          marginal zone B cell differentiation
          type B pancreatic cell development
          Notch signaling pathway
          Notch receptor processing
          determination of left/right symmetry
          compartment pattern specification
          regulation of blood pressure
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          proximal/distal pattern formation
          astrocyte development
          regulation of somitogenesis
          spinal cord development
          cerebellar molecular layer formation
          cerebellar Purkinje cell layer structural organization
          hemopoiesis
          cell differentiation
          regulation of cell adhesion
          negative regulation of epithelial cell differentiation
          negative regulation of interleukin-10 production
          negative regulation of glial cell apoptotic process
          organ growth
          regulation of growth
          negative regulation of cell differentiation
          negative regulation of epidermal cell differentiation
          negative regulation of auditory receptor cell differentiation
          negative regulation of myeloid cell differentiation
          negative regulation of myoblast differentiation
          negative regulation of neuron differentiation
          positive regulation of Notch signaling pathway
          positive regulation of endocytosis
          positive regulation of transcription from RNA polymerase II promoter
          lateral inhibition
          skeletal muscle tissue growth
          regulation of skeletal muscle tissue growth
          positive regulation of skeletal muscle tissue growth
          neuron fate specification
          inner ear development
          regulation of neurogenesis
          regulation of cell division
          retina development in camera-type eye
          retina morphogenesis in camera-type eye
          Notch signaling pathway involved in arterial endothelial cell fate commitment
          left/right axis specification
          nephron development
          proximal tubule development
          loop of Henle development
          clathrin-mediated endocytosis
          endothelial tip cell fate specification
          neuronal stem cell population maintenance
          skin epidermis development
          regulation of vascular endothelial growth factor signaling pathway
          positive regulation of sprouting angiogenesis
          regulation of energy homeostasis
          Notch binding
          calcium ion binding
          protein binding
          Tat protein binding
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0