Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012168.5 | NP_036300.2 | 5 | 1158 | ||
GenBank mRNA | AF174594.1 | - | 4 | 550 | ||
AF187318.1 | - | 5 | 1006 | |||
AK308531.1 | - | 6 | 1140 | |||
AK313019.1 | - | 5 | 758 | |||
BC025233.1 | - | 5 | 742 | 5090925 | ||
BC096747.1 | - | 5 | 758 | 5259298 | ||
CR542110.1 | - | 5 | 667 |