Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC4A7 SIRNA Assays ›
Gene Symbol:
SLC4A7
Gene Name:
solute carrier family 4 member 7
Gene Aliases:
NBC2, NBC3, NBCN1, SBC2, SLC4A6
Chromosome Location:
Chr.3: 27372721 - 27484420 on Build GRCh38
UniGene ID:
Hs.250072
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
13389
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC4A7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258379.1
NP_001245308.1
5
737
NM_001258380.1
NP_001245309.1
5
499
NM_001321103.1
NP_001308032.1
5
749
NM_001321104.1
NP_001308033.1
5
749
NM_001321105.1
NP_001308034.1
5
749
NM_001321106.1
NP_001308035.1
5
737
NM_001321107.1
NP_001308036.1
5
737
NM_001321108.1
NP_001308037.1
5
749
NM_003615.4
NP_003606.3
5
499
NR_135541.1
5
737
NR_135542.1
5
737
NR_135543.1
5
737
NR_135544.1
5
737
XM_005265598.4
XP_005265655.1
5
1003
XM_005265600.4
XP_005265657.1
5
1004
XM_006713421.2
XP_006713484.1
5
737
XM_011534256.2
XP_011532558.1
5
1010
XM_011534258.1
XP_011532560.1
5
713
XM_011534261.2
XP_011532563.1
5
1014
XM_011534263.2
XP_011532565.1
5
713
XM_011534265.1
XP_011532567.1
5
713
XM_017007526.1
XP_016863015.1
6
634
XM_017007527.1
XP_016863016.1
5
737
GenBank mRNA
AB012130.1
-
2
185
AF047033.1
-
5
499
AF053755.1
-
2
177
AF089726.1
-
5
499
AK309734.1
-
5
737
EU499349.1
-
5
664
EU934246.1
-
5
664
EU934247.1
-
5
676
EU934248.1
-
5
676
EU934249.1
-
5
664
EU934250.1
-
5
676
FJ178574.1
-
5
664
FJ178575.1
-
5
664
FJ178576.1
-
5
664
GU354307.1
-
5
664
GU354308.1
-
5
676
GU354309.1
-
5
676
GU354310.1
-
5
676
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
inorganic anion transport
bicarbonate transport
sodium ion transmembrane transport
regulation of intracellular pH
auditory receptor cell development
anion transmembrane transport
inorganic anion exchanger activity
anion transmembrane transporter activity
sodium:bicarbonate symporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System