Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002031.2 | NP_002022.1 | 8 | 2157 | ||
XM_005266880.4 | XP_005266937.1 | 9 | 2229 | |||
XM_005266881.2 | XP_005266938.1 | 9 | 2146 | |||
XM_005266882.4 | XP_005266939.1 | 10 | 2458 | |||
XM_011535653.2 | XP_011533955.1 | 10 | 2461 | |||
XM_011535654.2 | XP_011533956.1 | 9 | 2243 | |||
XM_011535655.2 | XP_011533957.1 | 9 | 2240 | |||
XM_011535656.2 | XP_011533958.1 | 8 | 1614 | |||
XM_017010645.1 | XP_016866134.1 | 9 | 2347 | |||
GenBank mRNA | BC012916.1 | - | 8 | 2002 | 4515877 | |
U00803.1 | - | 8 | 2157 | |||
U22322.1 | - | 8 | 2122 |