Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS6KC1 SIRNA Assays ›
          Gene Symbol:
          RPS6KC1
          Gene Name:
          ribosomal protein S6 kinase C1
          Gene Aliases:
          RPK118, RSKL1, S6K-delta-1, S6PKh1, humS6PKh1
          Chromosome Location:
          Chr.1: 213051233 - 213378753 on Build GRCh38
          UniGene ID:
          Hs.591416
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 133955

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RPS6KC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136138.2 NP_001129610.1 2, 3 394
          NM_001287218.1 NP_001274147.1 3, 4 430
          NM_001287219.1 NP_001274148.1 4, 5 547
          NM_001287220.1 NP_001274149.1 3, 4 430
          NM_012424.4 NP_036556.2 3, 4 430
          XM_005273095.1 XP_005273152.1 3, 4 417
          XM_005273096.1 XP_005273153.1 4, 5 528
          XM_006711272.1 XP_006711335.1 3, 4 417
          XM_017001024.1 XP_016856513.1 1, 2 91
          XM_017001025.1 XP_016856514.1 3, 4 403
          XM_017001027.1 XP_016856516.1 4, 5 528
          XM_017001028.1 XP_016856517.1 4, 5 528
          XM_017001029.1 XP_016856518.1 4, 5 528
          XM_017001030.1 XP_016856519.1 4, 5 528
          XM_017001031.1 XP_016856520.1 4, 5 528
          XM_017001032.1 XP_016856521.1 3, 4 405
          XM_017001033.1 XP_016856522.1 4, 5 528
          XM_017001034.1 XP_016856523.1 4, 5 528
          GenBank mRNA AB070706.1 - 3, 4 258
          AF477978.1 - 3, 4 407
          AK122921.1 - 2, 3 365
          AK122989.1 - 3, 4 417
          AK295053.1 - 2, 3 365
          AK299298.1 - 4, 5 533
          BC104769.1 - 3, 4 344 8143772
          BC143473.1 - 3, 4 344
          BX647303.1 - 2, 3 166
          DB060692.1 - 3, 4 418

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          signal transduction
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          phosphatidylinositol binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-vtt26:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0