Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256182.1 | NP_001243111.1 | 9 | 1383 | ||
NM_001256183.1 | NP_001243112.1 | 8 | 1309 | |||
NM_013275.5 | NP_037407.4 | 8 | 1312 | |||
XM_006721181.1 | XP_006721244.1 | 8 | 1089 | |||
XM_006721184.2 | XP_006721247.1 | 7 | 1127 | |||
XM_011523051.2 | XP_011521353.1 | 9 | 1372 | |||
XM_011523053.2 | XP_011521355.1 | 9 | 1478 | |||
XM_011523054.2 | XP_011521356.1 | 9 | 1392 | |||
XM_011523055.2 | XP_011521357.1 | 10 | 1677 | |||
XM_011523056.2 | XP_011521358.1 | 9 | 1414 | |||
XM_011523057.2 | XP_011521359.1 | 8 | 1295 | |||
XM_017023182.1 | XP_016878671.1 | 8 | 1745 | |||
XM_017023183.1 | XP_016878672.1 | 9 | 1481 | |||
XM_017023184.1 | XP_016878673.1 | 9 | 1478 | |||
XM_017023185.1 | XP_016878674.1 | 7 | 1119 | |||
XM_017023186.1 | XP_016878675.1 | 11 | 1855 | |||
XM_017023187.1 | XP_016878676.1 | 7 | 1207 | |||
GenBank mRNA | AB209186.1 | - | 7 | 957 | ||
AF121775.1 | - | 8 | 1069 | |||
AK025964.1 | - | 5 | 680 | |||
AK290994.1 | - | 7 | 991 | |||
AY373756.1 | - | 8 | 1315 | |||
AY533563.1 | - | 9 | 1140 | |||
BC017437.1 | - | 8 | 1281 | |||
BC052290.1 | - | 8 | 1227 | |||
BC069013.1 | - | 8 | 1227 | |||
KM670019.1 | - | 910 | ||||
KM670020.1 | - | 910 |