Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013330.4 | NP_037462.1 | 3 | 395 | ||
NM_197972.2 | NP_932076.1 | 3 | 364 | |||
NR_104229.1 | 3 | 395 | ||||
XM_017001099.1 | XP_016856588.1 | 3 | 381 | |||
GenBank mRNA | AB209049.1 | - | 3 | 204 | ||
AF153191.2 | - | 3 | 234 | |||
AK094513.1 | - | 3 | 347 | |||
AK290701.1 | - | 3 | 229 | |||
AK301917.1 | - | 3 | 378 | |||
BC006983.1 | - | 3 | 226 | 3997152 | ||
CD107958.1 | - | 3 | 423 |