Search Thermo Fisher Scientific
Gene Symbol: PCOLCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013363.3 | NP_037495.1 | 4 | 796 | ||
GenBank mRNA | AF098269.1 | - | 4 | 522 | ||
AK075478.1 | - | 4 | 793 | |||
AK223633.1 | - | 4 | 718 | |||
AK315121.1 | - | 4 | 564 | |||
AY035400.1 | - | 4 | 685 | |||
AY358557.1 | - | 4 | 795 | |||
BC006265.2 | - | 4 | 549 | |||
CR536570.1 | - | 4 | 489 |