Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PCOLCE2 SIRNA Assays ›
          Gene Symbol:
          PCOLCE2
          Gene Name:
          procollagen C-endopeptidase enhancer 2
          Gene Aliases:
          PCPE2
          Chromosome Location:
          Chr.3: 142817860 - 142889203 on Build GRCh38
          UniGene ID:
          Hs.8944
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 134084

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PCOLCE2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013363.3 NP_037495.1 9 1530
          GenBank mRNA AF098269.1 - 9 1256
          AK075478.1 - 9 1527
          AK223633.1 - 9 1452
          AK315121.1 - 9 1298
          AY035400.1 - 9 1419
          AY358557.1 - 9 1529
          BC006265.2 - 9 1283
          CR536570.1 - 9 1223
          DQ173439.1 - 1 41
          DQ173440.1 - 1 43
          DQ173443.1 - 1 39
          DQ173448.1 - 1 29
          DQ173450.1 - 1 30
          DQ173451.1 - 1 29
          DQ173452.1 - 1 30
          DQ173454.1 - 1 41
          DQ173455.1 - 1 48
          DQ173456.1 - 1 42
          DQ173457.1 - 1 42
          DQ173460.1 - 1 42
          DQ173461.1 - 1 44
          DQ173462.1 - 1 40
          DQ173463.1 - 1 42
          DQ173464.1 - 1 42
          DQ173466.1 - 1 43
          DQ173467.1 - 1 48
          DQ173468.1 - 1 53
          DQ173471.1 - 1 32
          DQ173473.1 - 1 43
          DQ173474.1 - 1 37
          DQ173476.1 - 1 40
          DQ173477.1 - 1 41
          DQ173478.1 - 1 39
          DQ173479.1 - 1 49
          DQ173480.1 - 1 43
          DQ173481.1 - 1 24
          DQ173483.1 - 1 42
          DQ173484.1 - 1 38
          DQ173485.1 - 1 42
          DQ173486.1 - 1 42
          DQ173487.1 - 1 35
          DQ173488.1 - 1 41
          DQ173489.1 - 1 43
          DQ173491.1 - 1 39
          DQ173492.1 - 1 40

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of peptidase activity
          collagen binding
          heparin binding
          peptidase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-hpqzk:80/100.66.77.239:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0