Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNMA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282535.1 | NP_001269464.1 | 2 | 548 | ||
NM_013364.5 | NP_037496.4 | 2 | 548 | |||
GenBank mRNA | AF083116.1 | - | 1 | 239 | ||
AK312299.1 | - | 2 | 475 | |||
AL136878.1 | - | 2 | 440 | |||
BC105096.1 | - | 1 | 232 | 8144099 | ||
BC105098.1 | - | 1 | 232 | 8144101 | ||
DA237274.1 | - | 2 | 519 | |||
DR001205.1 | - | 2 | 548 |