Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135820.1 | NP_001129292.1 | 23 | 4353 | ||
NM_013390.2 | NP_037522.1 | 24 | 4542 | |||
XM_005251869.4 | XP_005251926.1 | 24 | 4175 | |||
GenBank mRNA | AB037833.2 | - | 24 | 4135 | ||
AF137030.1 | - | 24 | 4150 | |||
AK027818.1 | - | 5 | 662 | |||
AK075370.1 | - | 17 | 2270 | |||
AK300405.1 | - | 11 | 1993 | |||
AK303765.1 | - | 7 | 1041 | |||
BC044850.1 | - | 2 | 1734 | |||
BC140773.1 | - | 23 | 3855 | 9021641 | ||
BC144510.1 | - | 24 | 4010 | |||
BC146780.1 | - | 24 | 4135 | 9007233 | ||
BC171782.1 | - | 23 | 3856 |