Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321357.1 | NP_001308286.1 | 4 | 1134 | ||
NM_001321358.1 | NP_001308287.1 | 3 | 507 | |||
NM_001321359.1 | NP_001308288.1 | 3 | 502 | |||
NM_001321361.1 | NP_001308290.1 | 3 | 507 | |||
NM_001321362.1 | NP_001308291.1 | 3 | 507 | |||
NM_001321363.1 | NP_001308292.1 | 3 | 507 | |||
NM_013392.3 | NP_037524.1 | 3 | 502 | |||
GenBank mRNA | AF113249.1 | - | 3 | 414 | ||
AK001946.1 | - | 3 | 392 | |||
AK027538.1 | - | 3 | 410 | |||
AK122664.1 | - | 4 | 1134 | |||
AK223136.1 | - | 3 | 398 | |||
AK225540.1 | - | 3 | 428 | |||
AK301345.1 | - | 4 | 345 | |||
AL136682.1 | - | 3 | 389 | |||
BC001221.1 | - | 3 | 416 | 3534543 | ||
BQ949042.1 | - | 3 | 448 | |||
DA161015.1 | - | 3 | 508 |