Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBQLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013444.3 | NP_038472.2 | 1 | 943 | ||
GenBank mRNA | AB015344.1 | - | 1 | 14 | ||
AF189009.1 | - | 1 | 897 | |||
AF293385.1 | - | 1 | 662 | |||
AK297252.1 | - | 1 | 909 | |||
AK302885.1 | - | 1 | 909 | |||
AL442081.1 | - | 1 | 556 | |||
BC069237.1 | - | 2 | 814 | 4543266 |