Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014165.3 | NP_054884.1 | 3 | 493 | ||
GenBank mRNA | AF060508.1 | - | 3 | 439 | ||
AF161474.1 | - | 3 | 482 | |||
AK026245.1 | - | 2 | 338 | |||
AK311850.1 | - | 3 | 466 | |||
BC018802.1 | - | 3 | 479 | |||
BC039464.1 | - | 3 | 472 | 4388671 | ||
BP258367.1 | - | 1 | 28 | |||
DB253189.1 | - | 3 | 493 |