Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPP1R15A SIRNA Assays ›
          Gene Symbol:
          PPP1R15A
          Gene Name:
          protein phosphatase 1 regulatory subunit 15A
          Gene Aliases:
          GADD34
          Chromosome Location:
          Chr.19: 48872392 - 48876062 on Build GRCh38
          UniGene ID:
          Hs.631593
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 134242

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPP1R15A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_014330.3 NP_055145.3 2 790
          GenBank mRNA AK001361.1 - 2 761
          AK225632.1 - 2 761
          AK297615.1 - 3 678
          AK298765.1 - 2 75
          AK311366.1 - 2 315
          BC003067.2 - 2 743 3507556
          CR457259.1 - 1 521
          U83981.1 - 2 743

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          apoptotic process
          cellular response to DNA damage stimulus
          cell cycle arrest
          aging
          negative regulation of cell proliferation
          negative regulation of peptidyl-threonine phosphorylation
          negative regulation of protein processing
          endoplasmic reticulum unfolded protein response
          positive regulation of translational initiation in response to stress
          negative regulation of phosphoprotein phosphatase activity
          positive regulation of phosphoprotein phosphatase activity
          cellular response to UV
          response to endoplasmic reticulum stress
          negative regulation of protein dephosphorylation
          cellular response to drug
          response to immobilization stress
          regulation of translational initiation by eIF2 alpha dephosphorylation
          response to cocaine
          positive regulation of apoptotic process
          regulation of angiogenesis
          negative regulation of cell death
          regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          peptidyl-serine dephosphorylation
          protein localization to endoplasmic reticulum
          cellular response to antibiotic
          cellular response to carbohydrate stimulus
          cellular response to methyl methanesulfonate
          cellular response to calcium ion starvation
          response to environmental enrichment
          cellular response to oxygen-glucose deprivation
          positive regulation of peptidyl-serine dephosphorylation
          negative regulation of PERK-mediated unfolded protein response
          negative regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
          positive regulation of endoplasmic reticulum stress-induced eIF2 alpha dephosphorylation
          cellular response to desipramine
          cellular response to cordycepin
          cellular response to gold(3+)
          cellular response to methamphetamine hydrochloride
          negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
          protein binding
          protein phosphatase 1 binding
          protein phosphatase type 1 regulator activity
          protein kinase binding
          protein phosphatase type 1 activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mzbz4:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0