Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHITM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014394.2 | NP_055209.2 | 5 | 575 | ||
GenBank mRNA | AB009685.1 | - | 4 | 371 | ||
AF060923.1 | - | 5 | 449 | |||
AF078863.1 | - | 5 | 500 | |||
AF131820.1 | - | 5 | 502 | |||
AF161400.1 | - | 5 | 484 | |||
AK098770.1 | - | 6 | 388 | |||
AK130094.1 | - | 5 | 478 | |||
AK130097.1 | - | 5 | 443 | |||
AK225318.1 | - | 5 | 452 | |||
AK292864.1 | - | 5 | 443 | |||
AK298332.1 | - | 5 | 457 | |||
AK299937.1 | - | 4 | 481 | |||
AL136713.1 | - | 5 | 485 | |||
AY358824.1 | - | 5 | 450 | |||
BC010354.1 | - | 5 | 446 | 4093037 | ||
BC022872.1 | - | 5 | 457 | |||
CR533519.1 | - | 4 | 371 |