Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NCK2 SIRNA Assays ›
          Gene Symbol:
          NCK2
          Gene Name:
          NCK adaptor protein 2
          Gene Aliases:
          GRB4, NCKbeta
          Chromosome Location:
          Chr.2: 105744649 - 105894274 on Build GRCh38
          UniGene ID:
          Hs.529244
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 13457

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NCK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001004720.2 NP_001004720.1 4 1176
          NM_001004722.3 NP_001004722.1 4 564
          NM_003581.4 NP_003572.2 5 1286
          XM_006712797.3 XP_006712860.1 5 1942
          XM_011511991.2 XP_011510293.1 5 1380
          XM_011511992.2 XP_011510294.2 3 508
          XM_017005103.1 XP_016860592.1 6 1544
          XM_017005104.1 XP_016860593.1 5 1556
          XM_017005105.1 XP_016860594.1 5 1293
          GenBank mRNA AF043119.1 - 3 1038
          AF047487.1 - 4 1142
          BC000103.2 - 4 1060 3505427
          BC007195.1 - 4 1067 3608149
          KU178418.1 - 3 1010
          KU178419.1 - 2 288

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          immunological synapse formation
          actin filament organization
          signal transduction
          signal complex assembly
          epidermal growth factor receptor signaling pathway
          regulation of epidermal growth factor-activated receptor activity
          negative regulation of cell proliferation
          vesicle-mediated transport
          cell migration
          lamellipodium assembly
          positive regulation of actin filament polymerization
          negative regulation of peptidyl-serine phosphorylation
          positive regulation of translation in response to endoplasmic reticulum stress
          positive regulation of T cell proliferation
          T cell activation
          positive regulation of GTPase activity
          positive regulation of transcription from RNA polymerase II promoter
          vascular endothelial growth factor receptor signaling pathway
          ephrin receptor signaling pathway
          dendritic spine development
          positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          negative regulation of PERK-mediated unfolded protein response
          negative regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
          negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
          SH3/SH2 adaptor activity
          ARF guanyl-nucleotide exchange factor activity
          protein binding
          cytoskeletal adaptor activity
          receptor signaling complex scaffold activity
          protein complex binding
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0