Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3YL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282687.1 | NP_001269616.1 | 11 | 1076 | ||
NM_015677.3 | NP_056492.2 | 9 | 922 | |||
NR_104223.1 | 11 | 2481 | ||||
NR_104225.1 | 9 | 1117 | ||||
GenBank mRNA | AK026586.1 | - | 9 | 1117 | ||
AL050373.1 | - | 11 | 1089 | |||
BC008374.1 | - | 9 | 831 | 3996066 | ||
BC043234.1 | - | 11 | 2486 |