Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request:org.apache.struts2.dispatcher.StrutsRequestWrapper@523fa0c5
Request attributes: pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
Popular
Applications & Techniques
Shop All Products
Services
Support
Sign in
Quick Order
Search
Thermo Fisher Scientific
Search All
Search
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
Attention: Existing favorites lists will be migrated to Shared Lists by the end of 2020.
[Learn more about Shared Lists]
LOADING
Gene Symbol:
UCHL5
Gene Name:
ubiquitin C-terminal hydrolase L5
Gene Aliases:
CGI-70, INO80R, UCH-L5, UCH37
Chromosome Location:
Chr.1: 193012366 - 193060140 on Build GRCh38
UniGene ID:
Hs.145469
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
134743
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UCHL5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199261.1
NP_001186190.1
11
1205
NM_015984.3
NP_057068.1
11
1208
XM_005245244.4
XP_005245301.1
12
1369
XM_005245246.4
XP_005245303.1
12
1363
XM_005245247.4
XP_005245304.1
11
1143
XM_005245248.4
XP_005245305.1
11
1163
XM_005245249.4
XP_005245306.1
11
1062
XM_006711366.3
XP_006711429.1
12
1372
XM_006711367.3
XP_006711430.1
13
1447
XM_006711368.3
XP_006711431.1
12
3797
XM_006711370.3
XP_006711433.3
12
1325
XM_006711371.3
XP_006711434.1
12
1143
XM_011509607.2
XP_011507909.1
12
1183
XM_011509608.2
XP_011507910.1
13
1544
XM_011509609.2
XP_011507911.1
13
1448
XM_017001429.1
XP_016856918.1
13
1444
XM_017001431.1
XP_016856920.1
12
1366
XM_017001434.1
XP_016856923.1
12
1322
XM_017001435.1
XP_016856924.1
11
1241
XM_017001436.1
XP_016856925.1
12
1463
XM_017001437.1
XP_016856926.1
12
1460
XM_017001438.1
XP_016856927.1
12
1167
XM_017001439.1
XP_016856928.1
12
1585
XM_017001440.1
XP_016856929.1
11
1471
XM_017001441.1
XP_016856930.1
11
1444
XM_017001442.1
XP_016856931.1
11
1059
GenBank mRNA
AF147717.1
-
11
1206
AK024751.1
-
11
1151
AK225794.1
-
12
1291
AK296487.1
-
7
795
BC015521.1
-
11
1138
3912584
Back To Top
More Information
Panther Classification:
Molecular Function -
cysteine protease
hydrolase
protease
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
Display Info:
cookie.CK_ISO_CODE.value:us
cookie.CK_LANG_CODE.value:en
cookie.CK_DISPLAY_TYPE.value:
pageContext.request.scheme:http(not used)
syndicationProtocol:https
syndicationExtensionPrefix:
syndicationDisplayType:
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-2-prod-green:8080/10.253.226.220:8080.
git-commit: 0914912cb48844dc9118fffd0bcf8fc2c0fe1c71
git-url: http://victoria.invitrogen.com:8333/magellan/core.git
git-branch: origin/release/1.17.0-2021.02.06-1.0