Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318498.1 | NP_001305427.1 | 7 | 580 | ||
NM_001318499.1 | NP_001305428.1 | 7 | 645 | |||
NM_016055.5 | NP_057139.1 | 6 | 522 | |||
NR_134658.1 | 5 | 484 | ||||
NR_134659.1 | 8 | 775 | ||||
NR_134660.1 | 6 | 640 | ||||
XM_017017919.1 | XP_016873408.1 | 7 | 615 | |||
GenBank mRNA | AF151876.1 | - | 6 | 1078 | ||
AF161408.1 | - | 6 | 525 | |||
AK297746.1 | - | 7 | 580 | |||
AK309219.1 | - | 5 | 462 | |||
AK310005.1 | - | 8 | 754 | |||
BC009764.1 | - | 6 | 515 | 4093230 | ||
BI549412.1 | - | 5 | 476 | |||
BM817807.1 | - | 6 | 520 | |||
BM828996.1 | - | 6 | 521 | |||
BX354271.2 | - | 6 | 605 |