Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IRAK4 SIRNA Assays ›
Gene Symbol:
IRAK4
Gene Name:
interleukin 1 receptor associated kinase 4
Gene Aliases:
IPD1, IRAK-4, NY-REN-64, REN64
Chromosome Location:
Chr.12: 43758913 - 43789543 on Build GRCh38
UniGene ID:
Hs.138499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
134831
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IRAK4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114182.2
NP_001107654.1
9
1024
NM_001145256.1
NP_001138728.1
8
878
NM_001145257.1
NP_001138729.1
7
830
NM_001145258.1
NP_001138730.1
6
660
NM_016123.3
NP_057207.2
8
976
XM_005268943.3
XP_005269000.1
8
1081
XM_005268944.4
XP_005269001.1
8
1045
XM_005268945.4
XP_005269002.1
8
1289
XM_005268947.4
XP_005269004.1
9
1035
XM_005268948.2
XP_005269005.1
10
1075
XM_005268949.2
XP_005269006.1
7
935
XM_006719438.3
XP_006719501.1
9
1337
XM_006719439.2
XP_006719502.1
8
1442
XM_006719440.2
XP_006719503.1
8
853
XM_006719441.2
XP_006719504.1
7
738
XM_006719442.2
XP_006719505.1
8
742
XM_006719443.3
XP_006719506.1
7
885
XM_011538431.2
XP_011536733.1
9
1093
XM_011538432.1
XP_011536734.1
9
1129
XM_011538433.2
XP_011536735.1
8
1708
XM_017019390.1
XP_016874879.1
10
1210
XM_017019391.1
XP_016874880.1
9
1175
GenBank mRNA
AF155118.1
-
8
943
AF445802.1
-
7
894
AK000528.1
-
8
923
AK027301.2
-
8
1471
AK299944.1
-
7
827
AK304583.1
-
6
663
AK314288.1
-
8
965
AY283670.1
-
7
894
AY283671.1
-
7
892
AY340962.1
-
8
959
AY340963.1
-
9
1007
AY340964.1
-
7
813
AY340965.1
-
8
861
AY340966.1
-
9
1026
AY340967.1
-
10
1074
AY730682.1
-
7
894
BC013316.1
-
8
964
4287014
KU178631.1
-
7
894
KU178632.1
-
6
748
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cytokine production
toll-like receptor signaling pathway
neutrophil mediated immunity
MyD88-dependent toll-like receptor signaling pathway
JNK cascade
cytokine-mediated signaling pathway
toll-like receptor 9 signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
positive regulation of smooth muscle cell proliferation
neutrophil migration
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
interleukin-1 receptor binding
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0